Assay Details
| Gene Symbol: scamp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025525.1 | NP_001020696.1 | 3 - 4 | 256 | 79 | |
| GenBank mRNA | BC097198.1 | - | 3 - 4 | 256 | 79 | |
More Information
Set Membership: |
|

scamp4
Genomic Map