Assay Details
| Gene Symbol: slc8a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037102.1 | NP_001032179.1 | 1 - 2 | 1820 | 107 | |
| XM_005169366.4 | XP_005169423.1 | 2 - 3 | 1890 | 107 | ||
| XM_009306112.3 | XP_009304387.1 | 2 - 3 | 2012 | 107 | ||
| XM_021479613.1 | XP_021335288.1 | 2 - 3 | 2023 | 107 | ||
| XM_021479614.1 | XP_021335289.1 | 2 - 3 | 2012 | 107 | ||
| GenBank mRNA | AY934775.1 | - | 1 - 2 | 1820 | 107 | |
| BC095839.1 | - | 2 - 3 | 1920 | 107 | ||
| BC116515.1 | - | 2 - 3 | 1895 | 107 | ||
| JN016748.1 | - | 1 - 2 | 1820 | 107 | ||
More Information
Set Membership: |
|

slc8a1a
Genomic Map