Assay Details
| Gene Symbol: slc8a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037102.1 | NP_001032179.1 | 2 - 3 | 1925 | 84 | |
| XM_005169366.4 | XP_005169423.1 | 3 - 4 | 1995 | 84 | ||
| XM_009306112.3 | XP_009304387.1 | 3 - 4 | 2117 | 84 | ||
| XM_021479613.1 | XP_021335288.1 | 3 - 4 | 2128 | 84 | ||
| XM_021479615.1 | XP_021335290.1 | 2 - 3 | 236 | 84 | ||
| GenBank mRNA | AY934775.1 | - | 2 - 3 | 1925 | 84 | |
| AY937230.1 | - | 3 - 4 | 2060 | 84 | ||
| BC095839.1 | - | 3 - 4 | 2025 | 84 | ||
| BC116515.1 | - | 3 - 4 | 2000 | 84 | ||
| JN016748.1 | - | 2 - 3 | 1925 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021479615.1 transcripts.
Set Membership: |
|

slc8a1a
Genomic Map