Assay Details
| Gene Symbol: slc8a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037102.1 | NP_001032179.1 | 6 - 7 | 2049 | 81 | |
| XM_005169366.4 | XP_005169423.1 | 6 - 7 | 2104 | 81 | ||
| XM_009306112.3 | XP_009304387.1 | 7 - 8 | 2241 | 81 | ||
| XM_021479613.1 | XP_021335288.1 | 7 - 8 | 2252 | 81 | ||
| XM_021479615.1 | XP_021335290.1 | 6 - 7 | 360 | 81 | ||
| GenBank mRNA | AY934775.1 | - | 6 - 7 | 2049 | 81 | |
| AY937230.1 | - | 6 - 7 | 2169 | 81 | ||
| JN016748.1 | - | 6 - 7 | 2049 | 81 | ||
More Information
Set Membership: |
|

slc8a1a
Genomic Map