Assay Details
| Gene Symbol: slc8a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037102.1 | NP_001032179.1 | 8 - 9 | 2271 | 63 | |
| XM_005169366.4 | XP_005169423.1 | 8 - 9 | 2326 | 63 | ||
| XM_009306112.3 | XP_009304387.1 | 9 - 10 | 2463 | 63 | ||
| XM_021479613.1 | XP_021335288.1 | 9 - 10 | 2474 | 63 | ||
| XM_021479614.1 | XP_021335289.1 | 5 - 6 | 2340 | 63 | ||
| XM_021479615.1 | XP_021335290.1 | 8 - 9 | 582 | 63 | ||
| GenBank mRNA | AY934775.1 | - | 8 - 9 | 2271 | 63 | |
| JN016748.1 | - | 8 - 9 | 2271 | 63 | ||
More Information
Set Membership: |
|

slc8a1a
Genomic Map