Assay Details
| Gene Symbol: epas1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039806.2 | NP_001034895.2 | 11 - 12 | 1832 | 127 | |
| GenBank mRNA | BC163440.1 | - | 11 - 12 | 1609 | 127 | |
| DQ375242.1 | - | 11 - 12 | 1786 | 127 | ||
More Information
Set Membership: |
|

epas1b
Genomic Map