Assay Details
| Gene Symbol: epas1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039806.2 | NP_001034895.2 | 3 - 4 | 668 | 106 | |
| GenBank mRNA | BC163440.1 | - | 3 - 4 | 445 | 106 | |
| DQ375242.1 | - | 3 - 4 | 625 | 106 | ||
More Information
Set Membership: |
|

epas1b
Genomic Map