Assay Details
| Gene Symbol: epas1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039806.2 | NP_001034895.2 | 7 - 8 | 1166 | 63 | |
| GenBank mRNA | BC163440.1 | - | 7 - 8 | 943 | 63 | |
| DQ375242.1 | - | 7 - 8 | 1123 | 63 | ||
More Information
Set Membership: |
|

epas1b
Genomic Map