Assay Details
| Gene Symbol: slc2a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039808.1 | NP_001034897.1 | 3 - 4 | 504 | 69 | |
| GenBank mRNA | DQ152824.1 | - | 3 - 4 | 504 | 69 | |
More Information
Set Membership: |
|

slc2a1a
Genomic Map