Assay Details
| Gene Symbol: adora2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039813.2 | NP_001034902.1 | 1 - 2 | 371 | 88 | |
| XM_005168628.4 | XP_005168685.1 | 2 - 3 | 601 | 88 | ||
| GenBank mRNA | AY945802.1 | - | 1 - 2 | 349 | 88 | |
| BC163683.1 | - | 1 - 2 | 371 | 88 | ||
More Information
Set Membership: |
|

adora2b
Genomic Map