Assay Details
| Gene Symbol: tas1r2.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039831.1 | NP_001034920.1 | 5 - 6 | 1540 | 66 | |
| GenBank mRNA | AB200900.1 | - | 5 - 6 | 1540 | 66 | |
| AB289805.1 | - | 5 - 6 | 1540 | 66 | ||
More Information
Set Membership: |
|

tas1r2.1
Genomic Map