Assay Details
| Gene Symbol: klf5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039927.2 | NP_001035016.2 | 2 - 3 | 1070 | 78 | |
| GenBank mRNA | BC114290.1 | - | 2 - 3 | 1042 | 78 | |
| KC293898.1 | - | 2 - 3 | 1066 | 78 | ||
More Information
Set Membership: |
|

klf5l
Genomic Map