Assay Details
| Gene Symbol: slc26a3.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040175.1 | NP_001035265.1 | 17 - 18 | 2084 | 114 | |
| XM_005164755.4 | XP_005164812.1 | 17 - 18 | 2207 | 114 | ||
| GenBank mRNA | BC115355.1 | - | 17 - 18 | 2084 | 114 | |
More Information
Set Membership: |
|

slc26a3.2
Genomic Map