Assay Details
| Gene Symbol: rec8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040378.1 | NP_001035468.1 | 13 - 14 | 1418 | 61 | |
| GenBank mRNA | BC115312.1 | - | 13 - 14 | 1418 | 61 | |
More Information
Set Membership: |
|

rec8b
Genomic Map