Assay Details
| Gene Symbol: bcl11aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040391.1 | NP_001035481.1 | 1 - 2 | 164 | 57 | |
| GenBank mRNA | BC115319.1 | - | 1 - 2 | 164 | 57 | |
More Information
Set Membership: |
|

bcl11aa
Genomic Map