Assay Details
| Gene Symbol: nrd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044715.2 | NP_001038180.2 | 11 - 12 | 1117 | 64 | |
| XM_009296247.2 | XP_009294522.1 | 11 - 12 | 1174 | 64 | ||
| XM_009296248.2 | XP_009294523.1 | 11 - 12 | 1196 | 64 | ||
| XM_009296249.3 | XP_009294524.1 | 7 - 8 | 557 | 64 | ||
| GenBank mRNA | BC134859.1 | - | 11 - 12 | 1144 | 64 | |
More Information
Set Membership: |
|

nrd1b
Genomic Map