Assay Details
| Gene Symbol: nrd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044715.2 | NP_001038180.2 | 13 - 14 | 1313 | 79 | |
| XM_009296247.2 | XP_009294522.1 | 13 - 14 | 1370 | 79 | ||
| XM_009296248.2 | XP_009294523.1 | 13 - 14 | 1392 | 79 | ||
| XM_009296249.3 | XP_009294524.1 | 9 - 10 | 753 | 79 | ||
| GenBank mRNA | BC134859.1 | - | 13 - 14 | 1340 | 79 | |
More Information
Set Membership: |
|

nrd1b
Genomic Map