Assay Details
| Gene Symbol: nrd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044715.2 | NP_001038180.2 | 25 - 26 | 2536 | 77 | |
| XM_009296247.2 | XP_009294522.1 | 25 - 26 | 2581 | 77 | ||
| XM_009296248.2 | XP_009294523.1 | 25 - 26 | 2603 | 77 | ||
| XM_009296249.3 | XP_009294524.1 | 21 - 22 | 1964 | 77 | ||
| GenBank mRNA | BC134859.1 | - | 25 - 26 | 2534 | 77 | |
More Information
Set Membership: |
|

nrd1b
Genomic Map