Assay Details
| Gene Symbol: nrd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044715.2 | NP_001038180.2 | 6 - 7 | 692 | 115 | |
| XM_009296247.2 | XP_009294522.1 | 6 - 7 | 749 | 115 | ||
| XM_009296248.2 | XP_009294523.1 | 6 - 7 | 771 | 115 | ||
| XM_009296249.3 | XP_009294524.1 | 2 - 3 | 132 | 115 | ||
| GenBank mRNA | BC134859.1 | - | 6 - 7 | 719 | 115 | |
More Information
Set Membership: |
|

nrd1b
Genomic Map