Assay Details
| Gene Symbol: march2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044790.2 | NP_001038255.2 | 3 - 4 | 632 | 118 | |
| GenBank mRNA | BC133146.1 | - | 3 - 4 | 632 | 118 | |
More Information
Set Membership: |
|

march2
Genomic Map