Assay Details
| Gene Symbol: nrcama | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044805.1 | NP_001038270.1 | 18 - 19 | 2501 | 75 | |
| XM_005164636.4 | XP_005164693.1 | 22 - 23 | 2859 | 75 | ||
| XM_005164637.4 | XP_005164694.1 | 22 - 23 | 2860 | 75 | ||
| XM_005164638.4 | XP_005164695.1 | 22 - 23 | 2859 | 75 | ||
| XM_009300127.3 | XP_009298402.1 | 22 - 23 | 3013 | 75 | ||
| XM_009300128.3 | XP_009298403.1 | 21 - 22 | 2784 | 75 | ||
| XM_009300129.3 | XP_009298404.1 | 21 - 22 | 2841 | 75 | ||
| XM_009300130.3 | XP_009298405.1 | 21 - 22 | 2829 | 75 | ||
| XM_009300131.3 | XP_009298406.1 | 22 - 23 | 2859 | 75 | ||
| XM_009300132.3 | XP_009298407.1 | 21 - 22 | 2799 | 75 | ||
| XM_009300134.3 | XP_009298409.1 | 22 - 23 | 2859 | 75 | ||
| XM_009300137.3 | XP_009298412.1 | 22 - 23 | 2859 | 75 | ||
| XM_009300138.3 | XP_009298413.1 | 21 - 22 | 2829 | 75 | ||
| XM_009300139.3 | XP_009298414.1 | 20 - 21 | 2811 | 75 | ||
| XM_021475020.1 | XP_021330695.1 | 22 - 23 | 2859 | 75 | ||
| XM_021475021.1 | XP_021330696.1 | 17 - 18 | 2444 | 75 | ||
| XM_021475022.1 | XP_021330697.1 | 22 - 23 | 2860 | 75 | ||
| XM_021475023.1 | XP_021330698.1 | 17 - 18 | 2474 | 75 | ||
| GenBank mRNA | BC171560.1 | - | 17 - 18 | 2471 | 75 | |
More Information
Set Membership: |
|

nrcama
Genomic Map