Assay Details
| Gene Symbol: clk2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044879.2 | NP_001038344.1 | 9 - 10 | 1246 | 76 | |
| XM_005159437.4 | XP_005159494.1 | 8 - 9 | 1164 | 76 | ||
| XM_009294047.3 | XP_009292322.1 | 9 - 10 | 1267 | 76 | ||
| GenBank mRNA | BC057521.1 | - | 8 - 9 | 1098 | 76 | |
| BC171570.1 | - | 9 - 10 | 1166 | 76 | ||
| BC171572.1 | - | 9 - 10 | 1166 | 76 | ||
More Information
Set Membership: |
|

clk2a
Genomic Map