Assay Details
| Gene Symbol: scn4ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045065.1 | NP_001038530.1 | 11 - 12 | 1821 | 77 | |
| XM_005171585.4 | XP_005171642.1 | 12 - 13 | 2172 | 77 | ||
| GenBank mRNA | BC163558.1 | - | 11 - 12 | 1867 | 77 | |
| DQ149505.1 | - | 11 - 12 | 1821 | 77 | ||
More Information
Set Membership: |
|

scn4ab
Genomic Map