Assay Details
| Gene Symbol: si:dkey-153k10.9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045110.1 | NP_001038575.1 | 19 - 20 | 2685 | 65 | |
| XM_005164587.4 | XP_005164644.1 | 19 - 20 | 2761 | 65 | ||
| Gene Symbol: si:ch211-233h19.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | EB937877.1 | - | 3 - 4 | 251 | 65 | |
More Information
Set Membership: |
|

si:dkey-153k10.9
Genomic Map