Assay Details
| Gene Symbol: si:dkey-153k10.9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045110.1 | NP_001038575.1 | 21 - 22 | 3033 | 87 | |
| Gene Symbol: si:ch211-233h19.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | EB937877.1 | - | 5 - 6 | 599 | 87 | |
More Information
Set Membership: |
|

si:dkey-153k10.9
Genomic Map