Assay Details
| Gene Symbol: si:dkey-153k10.9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045110.1 | NP_001038575.1 | 2 - 3 | 313 | 92 | |
| XM_005164587.4 | XP_005164644.1 | 2 - 3 | 389 | 92 | ||
More Information
Set Membership: |
|

si:dkey-153k10.9
Genomic Map