Assay Details
| Gene Symbol: setd1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045134.2 | NP_001038599.2 | 15 - 16 | 5116 | 96 | |
| XM_005169219.4 | XP_005169276.1 | 14 - 15 | 5245 | 96 | ||
| XM_005169220.4 | XP_005169277.1 | 15 - 16 | 4837 | 96 | ||
| XM_009305666.3 | XP_009303941.1 | 14 - 15 | 5248 | 96 | ||
| XM_009305667.3 | XP_009303942.1 | 14 - 15 | 5063 | 96 | ||
| XM_009305669.3 | XP_009303944.1 | 15 - 16 | 4483 | 96 | ||
| GenBank mRNA | DQ851809.1 | - | 2 - 3 | 226 | 96 | |
More Information
Set Membership: |
|

setd1ba
Genomic Map