Assay Details
| Gene Symbol: taf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045274.1 | NP_001038739.1 | 7 - 8 | 1072 | 75 | |
| XM_005160890.4 | XP_005160947.1 | 8 - 9 | 75 | |||
| GenBank mRNA | BC116466.1 | - | 7 - 8 | 1072 | 75 | |
| BC165337.1 | - | 7 - 8 | 985 | 75 | ||
More Information
Set Membership: |
|

taf1a
Genomic Map