Assay Details
| Gene Symbol: ttll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045305.1 | NP_001038770.1 | 11 - 12 | 1428 | 86 | |
| XM_009302562.3 | XP_009300837.1 | 2 - 3 | 379 | 86 | ||
| GenBank mRNA | BC117656.1 | - | 11 - 12 | 1428 | 86 | |
More Information
Set Membership: |
|

ttll3
Genomic Map