Assay Details
| Gene Symbol: gabrr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045376.1 | NP_001038841.1 | 3 - 4 | 591 | 70 | |
| XM_017351674.2 | XP_017207163.1 | 4 - 5 | 822 | 70 | ||
| GenBank mRNA | BC122282.1 | - | 3 - 4 | 591 | 70 | |
| BC164465.1 | - | 1 - 2 | 250 | 70 | ||
More Information
Set Membership: |
|

gabrr2a
Genomic Map