Assay Details
| Gene Symbol: elnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048064.2 | NP_001041529.2 | 15 - 16 | 1597 | 72 | |
| XM_017352790.2 | XP_017208279.1 | 15 - 16 | 1662 | 72 | ||
| XM_017352791.2 | XP_017208280.1 | 15 - 16 | 1662 | 72 | ||
| XM_017352793.2 | XP_017208282.1 | 15 - 16 | 1662 | 72 | ||
| XM_017352794.2 | XP_017208283.1 | 14 - 15 | 1593 | 72 | ||
| XM_017352796.2 | XP_017208285.1 | 15 - 16 | 1662 | 72 | ||
| XM_021468860.1 | XP_021324535.1 | 15 - 16 | 1662 | 72 | ||
| XM_021468861.1 | XP_021324536.1 | 15 - 16 | 1662 | 72 | ||
| XM_021468862.1 | XP_021324537.1 | 15 - 16 | 1662 | 72 | ||
| GenBank mRNA | DQ523563.1 | - | 15 - 16 | 1597 | 72 | |
| MF118567.1 | - | 3 - 4 | 147 | 72 | ||
| MF118569.1 | - | 3 - 4 | 147 | 72 | ||
| MF118570.1 | - | 3 - 4 | 147 | 72 | ||
| MF118571.1 | - | 3 - 4 | 147 | 72 | ||
More Information
Set Membership: |
|

elnb
Genomic Map