Assay Details
| Gene Symbol: elnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048064.2 | NP_001041529.2 | 19 - 20 | 1973 | 86 | |
| XM_017352790.2 | XP_017208279.1 | 19 - 20 | 2038 | 86 | ||
| XM_017352791.2 | XP_017208280.1 | 19 - 20 | 2038 | 86 | ||
| XM_017352793.2 | XP_017208282.1 | 19 - 20 | 2038 | 86 | ||
| XM_017352794.2 | XP_017208283.1 | 18 - 19 | 1969 | 86 | ||
| XM_017352796.2 | XP_017208285.1 | 19 - 20 | 2038 | 86 | ||
| XM_021468860.1 | XP_021324535.1 | 19 - 20 | 2038 | 86 | ||
| XM_021468861.1 | XP_021324536.1 | 19 - 20 | 2038 | 86 | ||
| XM_021468862.1 | XP_021324537.1 | 19 - 20 | 2038 | 86 | ||
| GenBank mRNA | DQ523563.1 | - | 19 - 20 | 86 | ||
| MF118567.1 | - | 7 - 8 | 86 | |||
| MF118568.1 | - | 7 - 8 | 86 | |||
| MF118569.1 | - | 7 - 8 | 86 | |||
| MF118570.1 | - | 7 - 8 | 86 | |||
| MF118571.1 | - | 7 - 8 | 86 | |||
| MF118572.1 | - | 7 - 8 | 86 | |||
More Information
Set Membership: |
|

elnb
Genomic Map