Assay Details
| Gene Symbol: elnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048064.2 | NP_001041529.2 | 21 - 22 | 2323 | 103 | |
| XM_017352790.2 | XP_017208279.1 | 21 - 22 | 2388 | 103 | ||
| XM_017352791.2 | XP_017208280.1 | 21 - 22 | 2388 | 103 | ||
| XM_017352793.2 | XP_017208282.1 | 21 - 22 | 2388 | 103 | ||
| XM_017352794.2 | XP_017208283.1 | 20 - 21 | 2319 | 103 | ||
| XM_017352796.2 | XP_017208285.1 | 21 - 22 | 2388 | 103 | ||
| XM_021468860.1 | XP_021324535.1 | 21 - 22 | 2388 | 103 | ||
| XM_021468861.1 | XP_021324536.1 | 21 - 22 | 2388 | 103 | ||
| XM_021468862.1 | XP_021324537.1 | 21 - 22 | 2388 | 103 | ||
| GenBank mRNA | DQ523563.1 | - | 21 - 22 | 2323 | 103 | |
| MF118567.1 | - | 9 - 10 | 873 | 103 | ||
| MF118568.1 | - | 9 - 10 | 873 | 103 | ||
| MF118569.1 | - | 9 - 10 | 873 | 103 | ||
| MF118570.1 | - | 9 - 10 | 873 | 103 | ||
| MF118571.1 | - | 9 - 10 | 873 | 103 | ||
| MF118572.1 | - | 10 - 11 | 693 | 103 | ||
| MF118574.1 | - | 2 - 3 | 111 | 103 | ||
| MF118575.1 | - | 2 - 3 | 96 | 103 | ||
| MF118576.1 | - | 2 - 3 | 96 | 103 | ||
More Information
Set Membership: |
|

elnb
Genomic Map