Assay Details
| Gene Symbol: elnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048064.2 | NP_001041529.2 | 24 - 25 | 2929 | 69 | |
| XM_017352790.2 | XP_017208279.1 | 24 - 25 | 2994 | 69 | ||
| XM_017352791.2 | XP_017208280.1 | 24 - 25 | 2994 | 69 | ||
| XM_017352793.2 | XP_017208282.1 | 24 - 25 | 2994 | 69 | ||
| XM_017352794.2 | XP_017208283.1 | 23 - 24 | 2925 | 69 | ||
| XM_021468860.1 | XP_021324535.1 | 24 - 25 | 2994 | 69 | ||
| GenBank mRNA | DQ523563.1 | - | 24 - 25 | 2956 | 69 | |
| MF118573.1 | - | 1 - 2 | 130 | 69 | ||
| MF118575.1 | - | 5 - 6 | 729 | 69 | ||
| MF118577.1 | - | 4 - 5 | 655 | 69 | ||
More Information
Set Membership: |
|

elnb
Genomic Map