Assay Details
| Gene Symbol: elnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048064.2 | NP_001041529.2 | 29 - 30 | 3448 | 67 | |
| XM_017352790.2 | XP_017208279.1 | 29 - 30 | 3513 | 67 | ||
| XM_017352791.2 | XP_017208280.1 | 29 - 30 | 3513 | 67 | ||
| XM_017352793.2 | XP_017208282.1 | 29 - 30 | 3513 | 67 | ||
| XM_017352794.2 | XP_017208283.1 | 28 - 29 | 3444 | 67 | ||
| XM_017352796.2 | XP_017208285.1 | 25 - 26 | 2877 | 67 | ||
| XM_021468860.1 | XP_021324535.1 | 29 - 30 | 3513 | 67 | ||
| XM_021468861.1 | XP_021324536.1 | 27 - 28 | 3258 | 67 | ||
| GenBank mRNA | DQ523563.1 | - | 29 - 30 | 3475 | 67 | |
| MF118573.1 | - | 6 - 7 | 649 | 67 | ||
| MF118574.1 | - | 6 - 7 | 600 | 67 | ||
| MF118575.1 | - | 10 - 11 | 1248 | 67 | ||
| MF118576.1 | - | 9 - 10 | 840 | 67 | ||
| MF118577.1 | - | 9 - 10 | 1174 | 67 | ||
| MF118578.1 | - | 6 - 7 | 559 | 67 | ||
| MF118579.1 | - | 6 - 7 | 511 | 67 | ||
More Information
Set Membership: |
|

elnb
Genomic Map