Assay Details
| Gene Symbol: elnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048064.2 | NP_001041529.2 | 2 - 3 | 306 | 76 | |
| XM_017352790.2 | XP_017208279.1 | 2 - 3 | 371 | 76 | ||
| XM_017352791.2 | XP_017208280.1 | 2 - 3 | 371 | 76 | ||
| XM_017352793.2 | XP_017208282.1 | 2 - 3 | 371 | 76 | ||
| XM_017352794.2 | XP_017208283.1 | 2 - 3 | 371 | 76 | ||
| XM_017352796.2 | XP_017208285.1 | 2 - 3 | 371 | 76 | ||
| XM_021468860.1 | XP_021324535.1 | 2 - 3 | 371 | 76 | ||
| XM_021468861.1 | XP_021324536.1 | 2 - 3 | 371 | 76 | ||
| XM_021468862.1 | XP_021324537.1 | 2 - 3 | 371 | 76 | ||
| GenBank mRNA | DQ523563.1 | - | 2 - 3 | 306 | 76 | |
| MF101834.1 | - | 2 - 3 | 301 | 76 | ||
| MF101835.1 | - | 2 - 3 | 301 | 76 | ||
| MF101836.1 | - | 2 - 3 | 301 | 76 | ||
| MF101837.1 | - | 2 - 3 | 301 | 76 | ||
| MF101838.1 | - | 2 - 3 | 301 | 76 | ||
More Information
Set Membership: |
|

elnb
Genomic Map