Assay Details
| Gene Symbol: elnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ523563.1 | - | 31 - 32 | 3721 | 87 | |
| MF118581.1 | - | 2 - 3 | 256 | 87 | ||
| MF118582.1 | - | 2 - 3 | 256 | 87 | ||
More Information
Set Membership: |
|

elnb
Genomic Map