Assay Details
| Gene Symbol: elnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048064.2 | NP_001041529.2 | 37 - 38 | 4273 | 66 | |
| XM_017352790.2 | XP_017208279.1 | 37 - 38 | 4485 | 66 | ||
| XM_017352791.2 | XP_017208280.1 | 37 - 38 | 4485 | 66 | ||
| XM_017352793.2 | XP_017208282.1 | 37 - 38 | 4485 | 66 | ||
| XM_017352794.2 | XP_017208283.1 | 36 - 37 | 4416 | 66 | ||
| XM_017352796.2 | XP_017208285.1 | 33 - 34 | 3849 | 66 | ||
| XM_021468860.1 | XP_021324535.1 | 36 - 37 | 4305 | 66 | ||
| XM_021468861.1 | XP_021324536.1 | 35 - 36 | 4230 | 66 | ||
| XM_021468862.1 | XP_021324537.1 | 31 - 32 | 3609 | 66 | ||
| GenBank mRNA | DQ523563.1 | - | 37 - 38 | 4330 | 66 | |
| MF118581.1 | - | 8 - 9 | 982 | 66 | ||
| MF118582.1 | - | 8 - 9 | 982 | 66 | ||
| MF118583.1 | - | 7 - 8 | 802 | 66 | ||
| MF118584.1 | - | 7 - 8 | 802 | 66 | ||
More Information
Set Membership: |
|

elnb
Genomic Map