Assay Details
| Gene Symbol: elnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048064.2 | NP_001041529.2 | 39 - 40 | 4417 | 60 | |
| XM_017352790.2 | XP_017208279.1 | 39 - 40 | 4629 | 60 | ||
| XM_017352791.2 | XP_017208280.1 | 39 - 40 | 4629 | 60 | ||
| XM_017352793.2 | XP_017208282.1 | 39 - 40 | 4629 | 60 | ||
| XM_017352794.2 | XP_017208283.1 | 38 - 39 | 4560 | 60 | ||
| XM_017352796.2 | XP_017208285.1 | 35 - 36 | 3993 | 60 | ||
| XM_021468860.1 | XP_021324535.1 | 38 - 39 | 4449 | 60 | ||
| XM_021468861.1 | XP_021324536.1 | 37 - 38 | 4374 | 60 | ||
| XM_021468862.1 | XP_021324537.1 | 33 - 34 | 3753 | 60 | ||
| GenBank mRNA | DQ523563.1 | - | 39 - 40 | 4474 | 60 | |
| MF118581.1 | - | 10 - 11 | 1126 | 60 | ||
| MF118582.1 | - | 10 - 11 | 1126 | 60 | ||
| MF118583.1 | - | 9 - 10 | 946 | 60 | ||
| MF118584.1 | - | 9 - 10 | 946 | 60 | ||
| MF118585.1 | - | 2 - 3 | 121 | 60 | ||
| MF118586.1 | - | 2 - 3 | 121 | 60 | ||
| MF118587.1 | - | 2 - 3 | 121 | 60 | ||
| MF118588.1 | - | 2 - 3 | 121 | 60 | ||
| MF118589.1 | - | 2 - 3 | 121 | 60 | ||
More Information
Set Membership: |
|

elnb
Genomic Map