Assay Details
| Gene Symbol: elnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048064.2 | NP_001041529.2 | 40 - 41 | 4458 | 60 | |
| XM_017352790.2 | XP_017208279.1 | 40 - 41 | 4670 | 60 | ||
| XM_017352793.2 | XP_017208282.1 | 40 - 41 | 4670 | 60 | ||
| XM_017352794.2 | XP_017208283.1 | 39 - 40 | 4601 | 60 | ||
| XM_017352796.2 | XP_017208285.1 | 36 - 37 | 4034 | 60 | ||
| XM_021468860.1 | XP_021324535.1 | 39 - 40 | 4490 | 60 | ||
| XM_021468861.1 | XP_021324536.1 | 38 - 39 | 4415 | 60 | ||
| XM_021468862.1 | XP_021324537.1 | 34 - 35 | 3794 | 60 | ||
| GenBank mRNA | DQ523563.1 | - | 40 - 41 | 4515 | 60 | |
| MF118581.1 | - | 11 - 12 | 1167 | 60 | ||
| MF118582.1 | - | 11 - 12 | 1167 | 60 | ||
| MF118586.1 | - | 3 - 4 | 162 | 60 | ||
| MF118589.1 | - | 3 - 4 | 162 | 60 | ||
More Information
Set Membership: |
|

elnb
Genomic Map