Assay Details
| Gene Symbol: elnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048064.2 | NP_001041529.2 | 42 - 43 | 4519 | 97 | |
| XM_017352790.2 | XP_017208279.1 | 42 - 43 | 4731 | 97 | ||
| XM_017352793.2 | XP_017208282.1 | 42 - 43 | 4731 | 97 | ||
| XM_017352794.2 | XP_017208283.1 | 41 - 42 | 4662 | 97 | ||
| XM_017352796.2 | XP_017208285.1 | 38 - 39 | 4095 | 97 | ||
| XM_021468860.1 | XP_021324535.1 | 41 - 42 | 4551 | 97 | ||
| XM_021468861.1 | XP_021324536.1 | 40 - 41 | 4476 | 97 | ||
| XM_021468862.1 | XP_021324537.1 | 36 - 37 | 3855 | 97 | ||
| GenBank mRNA | DQ523563.1 | - | 42 - 43 | 4576 | 97 | |
| MF118586.1 | - | 5 - 6 | 223 | 97 | ||
| MF118589.1 | - | 5 - 6 | 223 | 97 | ||
More Information
Set Membership: |
|

elnb
Genomic Map