Assay Details
| Gene Symbol: elnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048064.2 | NP_001041529.2 | 45 - 46 | 4670 | 70 | |
| XM_017352790.2 | XP_017208279.1 | 45 - 46 | 4882 | 70 | ||
| XM_017352791.2 | XP_017208280.1 | 44 - 45 | 4855 | 70 | ||
| XM_017352793.2 | XP_017208282.1 | 45 - 46 | 4882 | 70 | ||
| XM_017352794.2 | XP_017208283.1 | 44 - 45 | 4813 | 70 | ||
| XM_017352796.2 | XP_017208285.1 | 41 - 42 | 4246 | 70 | ||
| XM_021468860.1 | XP_021324535.1 | 44 - 45 | 4702 | 70 | ||
| XM_021468861.1 | XP_021324536.1 | 43 - 44 | 4627 | 70 | ||
| XM_021468862.1 | XP_021324537.1 | 39 - 40 | 4006 | 70 | ||
| GenBank mRNA | DQ523563.1 | - | 45 - 46 | 4727 | 70 | |
| MF118585.1 | - | 7 - 8 | 347 | 70 | ||
| MF118587.1 | - | 7 - 8 | 347 | 70 | ||
| MF118588.1 | - | 7 - 8 | 347 | 70 | ||
| MF118589.1 | - | 8 - 9 | 374 | 70 | ||
More Information
Set Membership: |
|

elnb
Genomic Map