Assay Details
| Gene Symbol: elnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048064.2 | NP_001041529.2 | 47 - 48 | 5149 | 92 | |
| XM_017352790.2 | XP_017208279.1 | 46 - 47 | 5424 | 92 | ||
| XM_017352791.2 | XP_017208280.1 | 45 - 46 | 5397 | 92 | ||
| XM_017352793.2 | XP_017208282.1 | 46 - 47 | 5424 | 92 | ||
| XM_017352794.2 | XP_017208283.1 | 45 - 46 | 5355 | 92 | ||
| XM_017352796.2 | XP_017208285.1 | 42 - 43 | 4788 | 92 | ||
| XM_021468860.1 | XP_021324535.1 | 45 - 46 | 5244 | 92 | ||
| XM_021468861.1 | XP_021324536.1 | 44 - 45 | 5169 | 92 | ||
| XM_021468862.1 | XP_021324537.1 | 40 - 41 | 4548 | 92 | ||
| GenBank mRNA | DQ523563.1 | - | 47 - 48 | 5185 | 92 | |
| MF118590.1 | - | 2 - 3 | 467 | 92 | ||
| MF118591.1 | - | 2 - 3 | 278 | 92 | ||
| MF118593.1 | - | 2 - 3 | 467 | 92 | ||
| MF118594.1 | - | 2 - 3 | 467 | 92 | ||
| MF118595.1 | - | 3 - 4 | 425 | 92 | ||
| MF118596.1 | - | 2 - 3 | 467 | 92 | ||
More Information
Set Membership: |
|

elnb
Genomic Map