Assay Details
| Gene Symbol: elnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048064.2 | NP_001041529.2 | 49 - 50 | 5365 | 87 | |
| XM_017352790.2 | XP_017208279.1 | 48 - 49 | 5640 | 87 | ||
| XM_017352791.2 | XP_017208280.1 | 47 - 48 | 5613 | 87 | ||
| XM_017352793.2 | XP_017208282.1 | 48 - 49 | 5640 | 87 | ||
| XM_017352794.2 | XP_017208283.1 | 47 - 48 | 5571 | 87 | ||
| XM_017352796.2 | XP_017208285.1 | 44 - 45 | 5004 | 87 | ||
| XM_021468860.1 | XP_021324535.1 | 47 - 48 | 5460 | 87 | ||
| XM_021468861.1 | XP_021324536.1 | 46 - 47 | 5385 | 87 | ||
| XM_021468862.1 | XP_021324537.1 | 42 - 43 | 4764 | 87 | ||
| GenBank mRNA | DQ523563.1 | - | 49 - 50 | 5401 | 87 | |
| MF118597.1 | - | 1 - 2 | 39 | 87 | ||
| MF118598.1 | - | 1 - 2 | 39 | 87 | ||
| MF118599.1 | - | 1 - 2 | 39 | 87 | ||
| MF118600.1 | - | 1 - 2 | 39 | 87 | ||
| MF118601.1 | - | 1 - 2 | 39 | 87 | ||
| MF118602.1 | - | 1 - 2 | 39 | 87 | ||
More Information
Set Membership: |
|

elnb
Genomic Map