Assay Details
| Gene Symbol: elnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048064.2 | NP_001041529.2 | 50 - 51 | 5409 | 58 | |
| XM_017352790.2 | XP_017208279.1 | 49 - 50 | 5684 | 58 | ||
| XM_017352791.2 | XP_017208280.1 | 48 - 49 | 5657 | 58 | ||
| XM_017352793.2 | XP_017208282.1 | 49 - 50 | 5684 | 58 | ||
| XM_017352794.2 | XP_017208283.1 | 48 - 49 | 5615 | 58 | ||
| XM_017352796.2 | XP_017208285.1 | 45 - 46 | 5048 | 58 | ||
| XM_021468860.1 | XP_021324535.1 | 48 - 49 | 5504 | 58 | ||
| XM_021468861.1 | XP_021324536.1 | 47 - 48 | 5429 | 58 | ||
| XM_021468862.1 | XP_021324537.1 | 43 - 44 | 4808 | 58 | ||
| GenBank mRNA | DQ523563.1 | - | 50 - 51 | 5445 | 58 | |
| MF118597.1 | - | 2 - 3 | 83 | 58 | ||
| MF118598.1 | - | 2 - 3 | 83 | 58 | ||
| MF118599.1 | - | 2 - 3 | 83 | 58 | ||
| MF118600.1 | - | 2 - 3 | 83 | 58 | ||
| MF118601.1 | - | 2 - 3 | 83 | 58 | ||
| MF118602.1 | - | 2 - 3 | 83 | 58 | ||
More Information
Set Membership: |
|

elnb
Genomic Map