Assay Details
| Gene Symbol: elnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048064.2 | NP_001041529.2 | 58 - 59 | 6025 | 72 | |
| XM_017352790.2 | XP_017208279.1 | 58 - 59 | 6342 | 72 | ||
| XM_017352791.2 | XP_017208280.1 | 57 - 58 | 6315 | 72 | ||
| XM_017352793.2 | XP_017208282.1 | 57 - 58 | 6300 | 72 | ||
| XM_017352794.2 | XP_017208283.1 | 57 - 58 | 6273 | 72 | ||
| XM_017352796.2 | XP_017208285.1 | 54 - 55 | 5706 | 72 | ||
| XM_021468860.1 | XP_021324535.1 | 57 - 58 | 6162 | 72 | ||
| XM_021468861.1 | XP_021324536.1 | 56 - 57 | 6087 | 72 | ||
| XM_021468862.1 | XP_021324537.1 | 52 - 53 | 5466 | 72 | ||
| GenBank mRNA | DQ523563.1 | - | 58 - 59 | 6127 | 72 | |
| MF118597.1 | - | 10 - 11 | 741 | 72 | ||
| MF118598.1 | - | 10 - 11 | 684 | 72 | ||
| MF118599.1 | - | 10 - 11 | 741 | 72 | ||
| MF118600.1 | - | 10 - 11 | 741 | 72 | ||
| MF118601.1 | - | 10 - 11 | 741 | 72 | ||
| MF118602.1 | - | 10 - 11 | 684 | 72 | ||
More Information
Set Membership: |
|

elnb
Genomic Map