Assay Details
| Gene Symbol: apoda.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076582.2 | NP_001070050.2 | 3 - 4 | 265 | 68 | |
| GenBank mRNA | BC122342.1 | - | 3 - 4 | 265 | 68 | |
| EH475543.1 | - | 2 - 3 | 588 | 68 | ||
More Information
Set Membership: |
|

apoda.1
Genomic Map