Assay Details
| Gene Symbol: myh7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077464.2 | NP_001070932.2 | 33 - 34 | 5057 | 111 | |
| GenBank mRNA | BC124103.1 | - | 1 - 2 | 103 | 111 | |
| BC150349.1 | - | 3 - 4 | 458 | 111 | ||
| BC165453.1 | - | 1 - 2 | 135 | 111 | ||
More Information
Set Membership: |
|

myh7l
Genomic Map