Assay Details
| Gene Symbol: slc20a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077546.1 | NP_001071014.1 | 6 - 7 | 1171 | 67 | |
| XM_005167168.4 | XP_005167225.1 | 6 - 7 | 1236 | 67 | ||
| XM_005167169.4 | XP_005167226.1 | 6 - 7 | 1298 | 67 | ||
| XM_005167171.4 | XP_005167228.1 | 6 - 7 | 1236 | 67 | ||
| XM_021478264.1 | XP_021333939.1 | 6 - 7 | 1071 | 67 | ||
| GenBank mRNA | BC124206.1 | - | 6 - 7 | 1171 | 67 | |
| BC165617.1 | - | 5 - 6 | 766 | 67 | ||
More Information
Set Membership: |
|

slc20a2
Genomic Map