Assay Details
| Gene Symbol: eno4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077547.1 | NP_001071015.1 | 2 - 3 | 402 | 69 | |
| XM_005158694.4 | XP_005158751.1 | 2 - 3 | 554 | 69 | ||
| XM_021467182.1 | XP_021322857.1 | 2 - 3 | 349 | 69 | ||
| GenBank mRNA | BC124782.1 | - | 2 - 3 | 402 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021467182.1 transcripts.
Set Membership: |
|

eno4
Genomic Map